Learn More
Potassium hydroxide, 50% w/v aq. soln., Thermo Scientific Chemicals
Description
Potassium hydroxide acts as the precursor for most of the soft and liquid soaps, and for numerous potassium-containing chemicals. It is widely used in the manufacture of detergents, fertilizer, rayon, paper, dyestuffs, various drugs, drug intermediates, fine chemicals, and petroleum products. In the field of organic chemistry, potassium hydroxide acts as a base, and is used in the synthesis of several organic compounds. Further it finds application as an electrolyte in alkaline batteries, which are based on nickel-cadmium, nickel-hydrogen and manganese dioxide-zinc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Colorless |
Density | 1.5 g/cm3 at 20°C |
Solubility Information | Miscible with water and ethanol. |
Quantity | 4 L |
Formula Weight | 56.11 |
Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
Physical Form | Liquid |
Chemical Name or Material | Potassium hydroxide |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N26L514 | Certificate of Analysis | 26/03/2025 | 035621.AP,035621.K2,035621.K7 |
62500029 | Certificate of Analysis | 03/02/2025 | 035621.AP,035621.K2,035621.K7 |
M11L513 | Certificate of Analysis | 20/01/2025 | 035621.AP,035621.K2,035621.K7 |
X07K512 | Certificate of Analysis | 20/11/2024 | 035621.AP,035621.K2,035621.K7 |
X01K523 | Certificate of Analysis | 14/11/2024 | 035621.AP,035621.K2,035621.K7 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.