missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide, 0.1N Standardized Solution, Thermo Scientific Chemicals

£26.13 - £131.66

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
SMILES [OH-].[K+]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11333707
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035599.K2
1 L
£26.13
1 L
Please to purchase this item. Need a web account? Register with us today!
11323707
View Documents
Thermo Scientific Alfa Aesar
035599.K7
4 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11313707
View Documents
Thermo Scientific Alfa Aesar
035599.M4
20 L
£131.66
20 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potassium hydroxide, 0.1N Standardized Solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1310-58-3
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
[OH-].[K+]
HKO
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
CHEBI:32035

Specifications

1310-58-3
Liquid
MFCD00003553
Freely soluble in water. Soluble in ethanol.
[OH-].[K+]
56.11
CHEBI:32035
Colorless liquid
Potassium hydroxide
Colorless
HKO
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium;hydroxide
14797
56.11
23 hPa (17mm Hg) at 20°C
Videos
Safety and Handling

Safety and Handling

H315-H319: Causes skin irritation. Causes serious eye irritation.

missing translation for 'einecsNumber' : 215-181-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only