missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydrogen Diiodate, ≥99.8% (RT), Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Color | White |
Physical Form | Powder |
Quantity | 100 g |
Linear Formula | KH(IO3)2 |
UN Number | UN3085 |
Formula Weight | 389.91g/mol |
Percent Purity | ≥99.8% (RT) |
Packaging | Glass bottle |
Chemical Name or Material | Potassium hydrogen diiodate |
Safety and Handling


Product Identifier
- Potassium hydrogen diiodate
- Danger
- Oxidising solid Category 2
- Skin corrosion/irritation Category 1 B
- H272-May intensify fire; oxidiser.
- H314-Causes severe skin burns and eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Potassium hydrogen diiodate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium Hydrogen Diiodate, ≥99.8% (RT), Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction