missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen carbonate, 99%, Thermo Scientific Chemicals

£23.62 - £259.29

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
Synonym potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
PubChem CID 516893
ChEBI CHEBI:81862
IUPAC Name potassium;hydrogen carbonate
SMILES [K+].OC([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11449010
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12429.36
500 g
£23.62
500g
Please to purchase this item. Need a web account? Register with us today!
11439010
View Documents
Thermo Scientific Alfa Aesar
A12429.0E
2500 g
£74.57
2500g
Please to purchase this item. Need a web account? Register with us today!
11429010
View Documents
Thermo Scientific Alfa Aesar
A12429.0C
10,000 g
£259.29
10000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potassium hydrogen carbonate is used in foods to regulate pH, as a fire suppression agent (BC dry chemical) and as a buffering agent in medications. It acts as an additive in winemaking and is an active component in club soda. Furthermore, it finds use in crops for the neutralization of acidic soils. It is also used as a source of carbon dioxide for leavening in baking. It is utilized to inhibit the growth of aspergillus parasiticus in czapek's agar. It is employed as a catalyst in synthetic fiber polymerization and olefin dehydrogenation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hydrogen carbonate is used in foods to regulate pH, as a fire suppression agent (BC dry chemical) and as a buffering agent in medications. It acts as an additive in winemaking and is an active component in club soda. Furthermore, it finds use in crops for the neutralization of acidic soils. It is also used as a source of carbon dioxide for leavening in baking. It is utilized to inhibit the growth of aspergillus parasiticus in czapek′s agar. It is employed as a catalyst in synthetic fiber polymerization and olefin dehydrogenation.

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Incompatible with strong oxidizing agents and strong acids.
Specifications

Chemical Identifiers

298-14-6
100.11
TYJJADVDDVDEDZ-UHFFFAOYSA-M
516893
potassium;hydrogen carbonate
CHKO3
MFCD00011402
potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
CHEBI:81862
[K+].OC([O-])=O

Specifications

298-14-6
MFCD00011402
potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
TYJJADVDDVDEDZ-UHFFFAOYSA-M
potassium;hydrogen carbonate
516893
100.12
Odorless
2.17
CHKO3
14,7609
Soluble in water. Insoluble in alcohol.
[K+].OC([O-])=O
100.11
CHEBI:81862
99%
Hygroscopic
Potassium hydrogen carbonate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 206-059-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only