Learn More
Potassium fluoride, 99.99% (metals basis), Thermo Scientific Chemicals
£31.76 - £291.45
Chemical Identifiers
CAS | 7789-23-3 |
---|---|
Molecular Formula | FK |
Molecular Weight (g/mol) | 58.10 |
MDL Number | MFCD00011398 |
InChI Key | NROKBHXJSPEDAR-UHFFFAOYSA-M |
Synonym | potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568 |
PubChem CID | 522689 |
ChEBI | CHEBI:66872 |
SMILES | [F-].[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11371568
|
Thermo Scientific Alfa Aesar
010980.04 |
2 g |
£31.76
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11381568
|
Thermo Scientific Alfa Aesar
010980.09 |
10 g |
£86.33
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11361568
|
Thermo Scientific Alfa Aesar
010980.18 |
50 g |
£291.45
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium fluoride is used in organic chemistry as a fluorinating agent to replace other halide ions like chorine, bromine etc. It is used as a catalyst in the manufacture of polyurethanes. Further, it is widely used for etching of glass, in disinfectant formulations and manufacture of optical glass. It is used in metal finishing, batteries, coatings and photographic chemicals. It is utilized for the study of ion-specific swelling and de-swelling of ampholytic polymer gels as well as in the measurement of electronic polarizabilities of ions in polymers of alkali halides. It finds application in the electronic industry as a metal surface treatment product. It is used as a preservative, a food additive, a catalyzer and a water absorbing agent. In the Finkelstein reaction, it is actively involved in the conversion of chlorocarbons to fluorocarbons using polar solvents like dimethyl formamide and ethylene glycol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7789-23-3 | |
58.10 | |
NROKBHXJSPEDAR-UHFFFAOYSA-M | |
522689 | |
[F-].[K+] |
FK | |
MFCD00011398 | |
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568 | |
CHEBI:66872 |
Specifications
1505°C | |
7789-23-3 | |
99.99% (metals basis) | |
MFCD00011398 | |
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568 | |
NROKBHXJSPEDAR-UHFFFAOYSA-M | |
potassium fluoride | |
522689 | |
58.1 | |
Hygroscopic | |
Potassium fluoride |
859°C | |
Crystalline | |
FK | |
3902818 | |
Freely soluble in water. Insoluble in alcohol. | |
[F-].[K+] | |
58.10 | |
CHEBI:66872 | |
1.363 | |
2.48 g/mL |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P311-P312-P330-P361-P363-P501c
H301+H311+H331-H318
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 232-151-5
missing translation for 'rtecsNumber' : TT0700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only