missing translation for 'onlineSavingsMsg'
Learn More

Potassium chromate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™

Product Code. 15603330
Click to view available options
Quantity:
250 g
1 kg
50 kg
6 x 1 kg
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-00-6
Molecular Formula CrK2O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00011368
InChI Key XMXNVYPJWBTAHN-UHFFFAOYSA-N
Synonym potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687
PubChem CID 24597
ChEBI CHEBI:75249
SMILES [K+].[K+].[O-][Cr]([O-])(=O)=O

Specifications

Quantity 1 kg
Linear Formula K2CrO4
UN Number 3288
IUPAC Name dipotassium;dioxido(dioxo)chromium
Formula Weight 194.19g/mol
Percent Purity ≥99.5%
Grade ACS/puriss. p.a.
Density Vapor Density: 6.7 (vs air)
Chemical Name or Material Potassium Chromate, Reag. Ph. Eur.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'