missing translation for 'onlineSavingsMsg'
Learn More
Potassium chloride, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E508, 99 to 100.5% (AT), ≤0.0001% Al, Honeywell Fluka™ Product Code.: 15611170

Potassium chloride, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E508, 99 to 100.5% (AT), ≤0.0001% Al, Honeywell Fluka™

Product Code. 15611170 Shop All Honeywell Fluka Products
250 g
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
6 x 2.5 kg
This item is not returnable. View return policy

Product Code. 15611170

Brand: Honeywell-Fluka 12636250G

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E508, 99-100.5% (AT), ≤0.0001% Al

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Quantity 250 g
Linear Formula KCl
UN Number NONH for all modes of transport
IUPAC Name potassium chloride
Formula Weight 74.55g/mol
Percent Purity 99 to 100.5% (AT)
Grade puriss.
Packaging Plastic Bottle
Density 1.98g/mL at 25°C
Chemical Name or Material Potassium Chloride, Meets analytical specification of Ph. Eur.BP, USP, FCC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium chloride, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E508, 99 to 100.5% (AT), ≤0.0001% Al, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.