missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Polytetrafluoroethylene Spinbar™ Flexible Magnetic Retrivers

Flexible magnetic Stirbar retriever recovers stir bars from lab sink drains and containers.

Brand:  Bel-Art™ H37774-0010

 View more versions of this product

Product Code. 12301288

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Flexible, Teflon™ rod with powerful Alnico Teflon™ coated magnet
  • Available in four colors, two magnet sizes
  • Flexible white rod is 33cm (13 in.) long
  • Autoclavable
Specifications

Specifications

Red
Retrieving magnetic stirring bars from containers or non-magnetic sink drains and traps
14.38in.
365 mm
Stir Bar Retriever
0.5in.
Stirring Bars
12.5 mm
Teflon Rod with Teflon Coated Alnico Magnet
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.