missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Polystyrene Petri Dishes

Manufactured from highly transparent polystyrene for its heat resistant properties to enable use with hot agar. Greiner BioOne™ Polystyrene Petri Dishes are available in a wide variety of formats and sizes to accommodate multiple research needs.

Brand:  Greiner Bio-One 688102

 View more versions of this product

Product Code. 10548071

  • £281.00 / Pack of 240

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Heat resistant up to 60°C
  • Available in vented and non-vented
  • Choose from standard round design, square-profile dish, special models and contact dishes/OneWell Plate™ models
  • Easy stacking
  • Crystal polystyrene for optimal analysis by microscopy
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Polystyrene
Non-sterile
Yes
120 mm
Single-use
Square
240 Pk.
17 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers