Learn More
Fisherbrand™ Polystyrene/Dacron Swab
Use these swabs for sample collection. Fisherbrand™ Polystyrene/Dacron Swab have a round, compact and shred resistant head and a snappable polystyrene shaft.
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 2.00000kg
Description
- Produced under strict ISO 9002/EN 29002 standards
- CE marked No. 98 06 0119, Class II A
Specifications
4000Pack | |
Snappable Polystyrene/Dacron | |
Snappable Polystyrene/Dacron | |
Snappable Polypropylene |
CE Certified | |
Swab | |
2.5mm | |
Dacron |