missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Multiroir™ Polypropylene Storage Bin with Handles
Store items in the Multiroir™ Polypropylene Storage Bin with Handles. The PP plastic and reliable handles provide a sturdy box in which to store many types of equipment, lab coats, instruments, and other medical and industrial supplies.
Specifications
Certifications/Compliance | 32128 Standard |
---|---|
Color | Transparent |
Material | Plastic |
Shape | Rectangular |
Includes | Handle |
Description
- Transparent polypropylene
- Good grip
- Orange handles
- Various capacities
Specifications
32128 Standard | |
Plastic | |
Handle | |
1 Each |
Transparent | |
Rectangular | |
Storage Box |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title