missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ark Plas Products™ Polypropylene Male Luers with Lock Ring
Use these luers with a wide variety of medical and laboratory instruments. Ark Plas Products™ Polypropylene Luer Plugs feature conical tapers molded to ISO 594-1 and ISO 594-2 Standards for 6% luer tapers to ensure compatibility with all other luers molded to the same standard.
Brand: Ark Plas Products™ LAX70-PP0
Description
- Natural polypropylene luer plugs
- Feature barbs designed with no parting line for reliable, leak-free use with a broad variety of medical and laboratory instruments, including syringetips and needles or stopcocks and needles
- Molded in virgin, lot-traceable material
- Ark-Plas Luer fittings include a large selection of both male and female luers available in locking or slip tip variances, as well as locking rings, bulkhead luers for panel mounting, threaded luers, adapters and plugs
Specifications
ISO 594-1 and 594-2 standards, RoHS Compliant, USP Class VI, FDA 21 CFR 177.1520, Animal Derivative-free, DEHP-free, BPA-free certified | |
1.6 mm | |
Flexible tubing or threaded ports | |
25/Pk. | |
Natural | |
Male Luer with Lock Ring |
Autoclavable | |
Polypropylene | |
-17°C | |
1/16 in. Barb | |
135°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction