Learn More
Masterflex™ Polypropylene Female Luer Fitting
Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Cole-Parmer™ Polypropylene Female Luer Fittings offer superior chemical resistance.
Brand:  Masterflex™ 30800-12
Product Code. 13598810
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Suited for ETO and gamma sterilization (not recommended for autoclaving)
  • Sold in packs of 25
Specifications
Specifications
Female Luer Fitting
Polypropylene
25 Pack