missing translation for 'onlineSavingsMsg'
Learn More
Polyethylene Glycol, Off-White Powder, MP Biomedicals™ Product Code.: 11435372
Add to basket

Polyethylene Glycol, Off-White Powder, MP Biomedicals™

Product Code. 11435372 Shop All MP Biomedicals Products
1 kg
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy

Product Code. 11435372

Brand: MP Biomedicals 0219544501

Add to basket
Add to basket
This item is not returnable. View return policy
Molecular Biology, Immunology
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym Carbowax, PEG
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Color White
Physical Form Powder
Density 1.1-1.2g/mL
Quantity 1 kg
Formula Weight 8000
Chemical Name or Material Polyethylene Glycol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Polyethylene Glycol, Off-White Powder, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.