missing translation for 'onlineSavingsMsg'
Learn More

Manutan Polycarbonate red door cabinet with one shelf, 915mm x 457mm x 457mm

Brand:  Manutan A343089

Code : Z3

Additional Details : Weight : 1.00000kg

Product Code. 12838219

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Strong welded construction. Floor Standing Cabinet
  • Red door
  • One shelf
  • Dimensions (H x W x D): 915mm x 457mm x 457mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers