missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(Vinyl Alcohol), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
Color | White to Yellow |
pH | 4.5 to 6.5 (4% aq. soln.) |
Physical Form | Powder |
Melting Point | 160.0°C to 240.0°C |
Ash | 90%% (Na2O) max. |
Linear Formula | [-CH2CH(OH)-]n |
Solubility Information | Solubility in water: soluble in hot water. |
Quantity | 500 g |
Percent Purity | 87.0 to 89.0% (degree of hydrolysis) |
Infrared Spectrum | Authentic |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction