missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PMS2L5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549101
Additional Details : Weight : 0.01000kg
Description
PMS2L5 Polyclonal antibody specifically detects PMS2L5 in Human samples. It is validated for Western Blot.Specifications
PMS2L5 | |
Polyclonal | |
Unconjugated | |
PMS2P5 | |
PMS1 homolog 2, mismatch repair system component pseudogene 5; PMS2L5; PMS2P5; PMS4; PMS7 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
5383 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
A8MQ11 | |
PMS2P5 | |
KLH conjugated synthetic peptide between 82-107 amino acids from the C-terminal region of human PMS2L5. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |