Learn More
PLEKHG3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566584
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: DSLSCQLSPEV DISVGVATED SPSVNGMEPP SPGCPVEPDR SSCKKKESAL STRDRLLLDK IKSYYENAEH HD Highest antigen sequence identity to the following orthologs - mouse 71%, rat 69%.
PLEKHG3 (Pleckstrin Homology And RhoGEF Domain Containing G3) is a Protein Coding gene. Diseases associated with PLEKHG3 include Congenital Hemolytic Anemia. Gene Ontology (GO) annotations related to this gene include Rho guanyl-nucleotide exchange factor activity. [GeneCards]Specifications
PLEKHG3 | |
Polyclonal | |
Unconjugated | |
PLEKHG3 | |
ARHGEF43; KIAA0599; PH domain-containing family G member 3; pleckstrin homology and RhoGEF domain containing G3; pleckstrin homology domain containing, family G (with RhoGef domain) member 3; pleckstrin homology domain containing, family G, member 3; pleckstrin homology domain-containing family G member 3; PLEKHG3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26030 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A1L390 | |
PLEKHG3 | |
Recombinant Human PLEKHG3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only