missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plasminogen Mouse anti-Human, Clone: 1F6A11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66399-1-IG-20UL
This item is not returnable.
View return policy
Description
Plasmin (PLG, Plasminogen) dissolves the fibrin of blood clots and acts as a proteolytic factor in a variety of other processes including embryonic development, tissue remodeling, tumor invasion, and inflammation. In ovulation, weakens the walls of the Graafian follicle. It activates the urokinase-type plasminogen activator, collagenases and several complement zymogens, such as C1 and C5. Cleavage of fibronectin and laminin leads to cell detachment and apoptosis. Also cleaves fibrin, thrombospondin and von Willebrand factor. Its role in tissue remodeling and tumor invasion may be modulated by CSPG4. Plasmin binds to cells.Specifications
Plasminogen | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P00747 | |
PLG | |
Plasminogen Fusion Protein Ag9938 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot, Immunohistochemistry (Paraffin) | |
1F6A11 | |
Unconjugated | |
PLG | |
Angiostatin, plasminogen, PLG | |
Mouse | |
Protein G | |
RUO | |
5340 | |
-20°C | |
Liquid |