missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PJA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA568182
Additional Details : Weight : 0.01000kg
Description
Has E2-dependent E3 ubiquitin-protein ligase activity. Ubiquitinates MAGED1 antigen leading to its subsequent degradation by proteasome (By similarity). May be involved in protein sorting.Specifications
PJA | |
Polyclonal | |
Unconjugated | |
PJA1 | |
RNF70 | |
Rabbit | |
Affinity chromatography | |
RUO | |
64219 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8NG27 | |
PJA1 | |
Synthesized peptide derived from human PJA. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |