missing translation for 'onlineSavingsMsg'
Learn More

Novacyt - Microgen Bioproducts Phosphate Buffered Saline

Microgen Bioproducts Phosphate Buffered Saline - pH 7.2, to make 10 L

Brand:  Novacyt - Microgen Bioproducts M34A

Product Code. 17383191

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Phosphate Buffered Saline (x 20 concentrate)
Specifications

Specifications

Clear
15 mM
Phosphate Buffered Saline
7.2
Dilute 1 in. 20 (500 mL to 10 L)
Phosphate Buffered Saline (PBS)
General Laboratory Use, and is Produced as a Convenient Means of Generating a Working Strength 15 mM Phosphate Buffered Saline.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.