missing translation for 'onlineSavingsMsg'
Learn More

PHF22 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA557394

Product Code. 15745291

  • £447.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Immunogen sequence: KRPADKMKSD ITEGVDIPKK PRLEKPETQS SPITVQSSKD LPMADLSSFE ETSADDFAME MGLACVVCRQ MMVASGNQLV ECQECHNLYH RDCH Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%.

INTS12 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit and mediates 3-prime end processing of small nuclear RNAs U1 and U2.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PHF22
Polyclonal
Unconjugated
Ints12
1110020M19Rik; 2810027J24Rik; 4930529N21Rik; A230056J18Rik; AI666778; hypothetical nuclear factor SBBI22; hypothetical protein LOC415251; int12; Integrator complex subunit 12; INTS12; PHD finger protein 22; PHF22; SBBI22; Unknown (protein for MGC:134302); wu:fi17f05; zgc:86602
Rabbit
Antigen affinity chromatography
RUO
57117
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry, Western Blot
0.5 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96CB8
Ints12
Recombinant protein corresponding to Human PHF22
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Research Use Only