missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein solution, Indicator, Reag. Ph. Eur., 1% in ethanol, Honeywell Fluka™

Product Code. 15644310
Click to view available options
Quantity:
30 mL
500 mL
1 L
6 x 500 mL
50 L
6 x 1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Indicator, Reag. Ph. Eur., 1% in ethanol

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-09-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00005913
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
PubChem CID 4764
ChEBI CHEBI:34914
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Linear Formula C20H14O4
Beilstein 284423
IUPAC Name 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
Grade Ph. Eur.
Packaging Poly Bottle
Quantity 6 x 500 mL
Chemical Name or Material Phenolphthalein solution, Indicator, Reag. Ph. Eur., 1% in ethanol
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenolphthalein solution
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : phenolphthalein, ethanol; ethyl alcohol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'