missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Phenol Red, sodium salt
£20.40 - £115.58
Chemical Identifiers
CAS | 34487-61-1 |
---|---|
Molecular Formula | C19H13NaO5S |
Molecular Weight (g/mol) | 376.358 |
InChI Key | HKHYOKBQJILTEI-UHFFFAOYSA-M |
PubChem CID | 23686673 |
IUPAC Name | sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate |
SMILES | C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10279223
|
Thermo Scientific Chemicals
151440050 |
5 g | Glass bottle |
£20.40
5g |
In Stock
Log in to see stock. |
||||
10418120
|
Thermo Scientific Chemicals
151440250 |
25 g | Glass bottle |
£42.91
25g |
In Stock
Log in to see stock. |
||||
11986801
|
Thermo Scientific Chemicals
151441000 |
100 g | Glass bottle |
£115.58
100g |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
34487-61-1 | |
376.358 | |
23686673 | |
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+] |
C19H13NaO5S | |
HKHYOKBQJILTEI-UHFFFAOYSA-M | |
sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate |
Specifications
285.0°C | |
C19H13NaO5S | |
HKHYOKBQJILTEI-UHFFFAOYSA-M | |
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+] | |
376.358 | |
376.35 | |
1600 (558nm, in buffer) min. | |
Brown to Red | |
Phenol Red, sodium salt |
34487-61-1 | |
Solubility in water: soluble. | |
Authentic | |
sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate | |
23686673 | |
Pure | |
pH 6.8 (yellow) to pH 8.2 (red) | |
Amorphous Powder |
Safety and Handling
Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Phenol Red, sodium salt
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P362-Take off contaminated clothing.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title