Learn More
Chemical Identifiers
CAS | 143-74-8 |
---|---|
Molecular Formula | C19H14O5S |
Molecular Weight (g/mol) | 354.38 |
MDL Number | MFCD00003552 |
InChI Key | BELBBZDIHDAJOR-UHFFFAOYSA-N |
Synonym | Phenolsulfonephthalein |
PubChem CID | 4766 |
ChEBI | CHEBI:31991 |
SMILES | OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10035950
|
Thermo Scientific Chemicals
417240050 |
5 g | Glass bottle |
£19.70
5g |
In Stock
Log in to see stock. |
||||
10157750
|
Thermo Scientific Chemicals
417240100 |
10 g | Glass bottle |
£30.15
10g |
In Stock
Log in to see stock. |
||||
10299473
|
Thermo Scientific Chemicals
417241000 |
100 g | Glass bottle |
£198.99
100g |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
143-74-8 | |
354.38 | |
BELBBZDIHDAJOR-UHFFFAOYSA-N | |
4766 | |
OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 |
C19H14O5S | |
MFCD00003552 | |
Phenolsulfonephthalein | |
CHEBI:31991 |
Specifications
>300.0°C | |
passes test | |
MFCD00003552 | |
15, 7360 | |
Solubility in water: 0.77g/L. Other solubilities: soluble in ethanol | |
Authentic | |
3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione | |
4766 | |
354.38 | |
from pH 6.8 (yellow) to pH 8.2 (red) | |
Liquid After Melting |
143-74-8 | |
C19H14O5S | |
19, 91 | |
Phenolsulfonephthalein | |
BELBBZDIHDAJOR-UHFFFAOYSA-N | |
OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 | |
354.38 | |
CHEBI:31991 | |
ACS Reagent | |
Colorless to Yellow | |
Phenol Red, Pure |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
- Phenol Red
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only