Learn More
SI Analytics™ pH Combination Electrode
Ensure high measurement accuracy, stability, and long service life while maintaining high adaptability to your measurement tasks. SI Analytics™ pH Combination Electrode is robust, durable, and easy to clean.
Specifications
pH Range 0 to 14
Membrane Sphere
Diameter (Metric) 12 mm
View More Specs
Description
Description
  • Proven high-end electrodes for demanding measurement tasks
  • Double junction Silamid™ reference system for fast and stable acquiring of measured values and for longer electrode life
  • Utmost versatility of pH electrodes is achieved by a large selection of junctions, membrane glass types and shapes, shaft lengths and diameters, ground joints, plug connections and integrated temperature sensors
  • Each pH combination electrode comes with individual serial number and quality certificate
Specifications
Specifications
0 to 14
Sphere
12 mm
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
SI Analytics™ pH Combination Electrode