Learn More
pH Buffer Solution 7.00 at 25°C, Color-Coded, Mercury-Free, Reagecon
Yellow-colored solution for ease of use. Reagecon pH Buffer Solution 7.00 at 25°C, Color-Coded, Mercury-Free is tested and certified by an ISO17025 Accredited Test Method, is NIST traceable, and has guaranteed stability throughout its entire shelf life, even after opening the bottle.
Brand: Reagecon 107025
Code : NEW
Additional Details : Weight : 1.00710kg
Description
Yellow-colored solution for ease of use. Reagecon pH Buffer Solution 7.00 at 25°C, Color-Coded, Mercury-Free is tested and certified by an ISO17025 Accredited Test Method, is NIST traceable, and has guaranteed stability throughout its entire shelf life, even after opening the bottle.
- Color coded solution for easy visual identification
- Tested at 25°C to two decimal places, with an accuracy of ±0.01
- Certified by Reagecon ISO 17025 Accredited Test Method (INAB Ref: 264T)
- NIST traceable
Specifications
pH Buffer Solution | |
7558-79-4,7778-77-0,102-46-5,1934-21-0,7732-18-5 | |
0.989 | |
7 | |
1 L | |
Certified by Reagecon’s ISO 17025 Accredited Test Method (INAB Ref: 264T) | |
HNa2O4P | |
HDPE screwtop bottle, GL45 | |
23 hPa (17.3mm Hg) | |
BNIILDVGGAEEIG-UHFFFAOYSA-L | |
disodium hydrogen phosphate |
0°C | |
100°C | |
Yellow; tested at 25deg;C | |
Liquid | |
pH Buffer | |
Disodium hydrogen orthophosphate, potassium dihydrogen orthophosphate, Kathon WTE, Trisodium 5-hydroxy-1-(4-sulphophenyl)-4-(4-sulphophenylazo)pyrazole-3-carboxylate, Water | |
MFCD00003496 | |
Yes | |
Fully miscible in water | |
[Na+].[Na+].OP([O-])([O-])=O | |
141.96 |