missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH Buffer Solution 4.00 at 25°C, Reagecon
Click to view available options
Quantity:
1 L
Description
- Tested at 25°C to two decimal places, with an accuracy of ±0.01
- Certified by Reagecon ISO 17025 Accredited Test Method (INAB Ref: 264T)
- NIST traceable
Specifications
CAS | 877-24-7, 102-46-5, 3244-88-0, 7732-18-5 |
Melting Point | 0°C |
Boiling Point | 100°C |
CAS Max % | 0.025, 0.01, 0.001, 0.989 |
Color | Clear; tested at 25deg;C |
pH | 4 |
Physical Form | Liquid |
Quantity | 1 L |
Formulation | Potassium hydrogen phthalate, Kathon WTE, Acid fuchsin dye, Water |
Molecular Formula | C8H5KO4 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'