missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25081-1-AP-150UL
This item is not returnable.
View return policy
Description
Glycerol-3-phosphate phosphatase hydrolyzing glycerol-3-phosphate into glycerol. Thereby, regulates the cellular levels of glycerol-3-phosphate a metabolic intermediate of glucose, lipid and energy metabolism. Was also shown to have a 2-phosphoglycolate phosphatase activity and a tyrosine-protein phosphatase activity. However, their physiological relevance is unclear. In vitro, has also a phosphatase activity toward ADP, ATP, GDP and GTP.Specifications
PGP | |
Polyclonal | |
Unconjugated | |
PGP | |
PGP, PGPase, phosphoglycolate phosphatase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
283871, 67078 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A6NDG6, Q8CHP8 | |
PGP | |
PGP Fusion Protein Ag18623 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |