missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum Ether, puriss. p.a. ACS Reagent, Honeywell™
41.19 GBP valid until 2024-03-31
Use promo code "21663" to get your promotional price.
Use promo code "21663" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
5 L
195 L
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
UN Number | 1268 |
Grade | ACS/puriss. p.a. |
Density | 0.640 to 0.655g/mL at 20°C |
Quantity | 1 L |
Flash Point | <−30°C (<−22°F) |
Chemical Name or Material | Petroleum Ether, Reag. ISO |
Safety and Handling
Product Identifier
- Petroleum ether
- Danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Aspiration toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H411-Toxic to aquatic life with long lasting effects.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Hydrocarbons, C6, Isoalkanes, <5% n-Hexane
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'