missing translation for 'onlineSavingsMsg'
Learn More

Petroleum ether, low boiling point, Honeywell™

£29.27 - £161.01

Chemical Identifiers

CAS 101316-46-5
MDL Number MFCD00081849
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15637070
View Documents
Honeywell Riedel-de Haën
77399-500ML
500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15607070
View Documents
Honeywell Riedel-de Haën
77399-1L
1 L
£71.99
1 L
Please to purchase this item. Need a web account? Register with us today!
15617070
View Documents Promotion Details
Honeywell Riedel-de Haën
77399-2.5L
2.5 L
£161.01
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

101316-46-5
MFCD00081849

Specifications

101316-46-5
30°C to 40°C
MFCD00081849
Petroleum ether
0.63g/mL at 20°C
−49°C (−56.2°F)
UN1268
Videos
Safety and Handling

Safety and Handling

P210-P260-P280-P301 + P310-P370 + P378-P403 + P235

Danger

missing translation for 'einecsNumber' : 309-852-0

SDS
Documents

Documents

Product Certifications