missing translation for 'onlineSavingsMsg'
Learn More

Petroleum ether, ACS reagent, Honeywell Riedel-de Haën™

ACS reagent

£20.13 - £272.31

Chemical Identifiers

CAS 8030-30-6
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC
View More Specs

Products 6
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15661660
View Documents
Honeywell-Riedel-de Haen
184519-500ML
500 mL Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15631660
View Documents
Honeywell-Riedel-de Haen
184519-1L
1 L Glass Bottle
£43.97
1 L
Please to purchase this item. Need a web account? Register with us today!
15641660
View Documents
Honeywell-Riedel-de Haen
184519-2.5L
2.5 L Glass Bottle
£112.57
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15681660
View Documents
Honeywell-Riedel-de Haen
184519-6X500ML
6 x 500 mL Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15671660
View Documents
Honeywell-Riedel-de Haen
184519-6X1L
6 x 1 L Glass Bottle
£205.86
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15651660
View Documents
Honeywell-Riedel-de Haen
184519-20L
20 L Stainless Steel Can This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
Specifications

Chemical Identifiers

8030-30-6
72.15
OFBQJSOFQDEBGM-UHFFFAOYSA-N
8003
pentane
C5H12
MFCD00009498
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
CHEBI:37830
CCCCC

Specifications

8030-30-6
<50.0000%
40°C to 60°C
C5H12
UN3295
OFBQJSOFQDEBGM-UHFFFAOYSA-N
pentane
8003
ACS Reagent
≥25.0000%
0.64 g/mL at 25 °C(lit.)
−30°C
MFCD00009498
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
CCCCC
72.15
CHEBI:37830
Petroleum ether, ACS reagent
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 1260 days from date of manufacture

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Hydrocarbons, liquid, n.o.s.

missing translation for 'einecsNumber' : 232-453-7

Recommended Storage : Room Temp

SDS
Documents

Documents

Product Certifications