Learn More
Thermo Scientific Chemicals Pepsin from Porcine Stomach
Derived from Porcine stomach
Brand:  Thermo Scientific Chemicals J20015.A1
Additional Details : CAS Number : 9001-75-6 Weight : 1.00000kg
 View more versions of this product
Product Code. 15845328
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • One unit causes an increase in absorbance at 280nm of 0.001/min. at 37°C at pH 2.0.
Specifications
Specifications
9001-75-6
1 kg
Pepsin
Powder
EC 3.4.23.1,Pepsin A
Product Suggestions
Product Suggestions
Videos
SDS Documents