missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentane, ULTRA RESI-ANALYZED™, J.T. Baker™
High purity solvents and salts developed for organic residue extraction/concentration procedures
£213.00 - £580.00
Chemical Identifiers
CAS | 109-66-0 |
---|---|
Molecular Formula | C5H12 |
Molecular Weight (g/mol) | 72.15 |
MDL Number | MFCD00009498 |
InChI Key | OFBQJSOFQDEBGM-UHFFFAOYSA-N |
Synonym | n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish |
PubChem CID | 8003 |
ChEBI | CHEBI:37830 |
IUPAC Name | pentane |
SMILES | CCCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15588574
|
Avantor J.T.Baker
9333-02 |
1 L | Narrow Mouth Amber Glass Bottle | N/A | |||||
15574055
|
Avantor J.T.Baker
9333-03 |
4 L | Narrow Mouth Amber Glass Bottle |
£580.00
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For use in Gas Chromatography (GC), Organic Residue Analysis; For Laboratory, Research, or Manufacturing Use
Chemical Identifiers
109-66-0 | |
72.15 | |
OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
8003 | |
pentane |
C5H12 | |
MFCD00009498 | |
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish | |
CHEBI:37830 | |
CCCCC |
Specifications
109-66-0 | |
C5H12 | |
1L = 0.63kg | |
OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
pentane | |
8003 | |
72.15 | |
Pentane |
−40.00°C(-40°F) | |
MFCD00009498 | |
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish | |
CCCCC | |
72.15 | |
CHEBI:37830 | |
ULTRA RESI-ANALYZED™ |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H336-May cause drowsiness or dizziness.
- H411-Toxic to aquatic life with long lasting effects.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title