missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Penicillin G sodium salt, 96+%, Thermo Scientific Chemicals Product Code.: 15525166

Thermo Scientific Chemicals Penicillin G sodium salt, 96+%, Thermo Scientific Chemicals

Product Code. 15525166
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15525166

Brand: Thermo Scientific Chemicals 455690010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Penicillin G sodium salt, CAS # 69-57-8, also called benzylpenicillin, is a β-lactam antibiotic active against gram-positive and gram-negative bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Penicillin G sodium salt is the sodium salt of benzylpenicillin, a broad-spectrum β-lactam antibiotic with gram-positive and gram-negative bactericidal activity
  • Penicillin G binds and inactivates penicillin-binding-proteins associated with the bacterial cell wall, thus inhibiting cell wall synthesis and resulting in cell lysis

Applications

  • Penicillin G can be used as a selection agent in the isolation of microorganisms or in cell culture applications
  • It can also be used to prevent infection in research animals
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-57-8
Molecular Formula C16H17N2NaO4S
Molecular Weight (g/mol) 356.372
InChI Key FCPVYOBCFFNJFS-LQDWTQKMSA-M
PubChem CID 23668834
ChEBI CHEBI:51765
IUPAC Name sodium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[Na+]

Specifications

Type Penicillin G Sodium Salt
pH 5.0 to 7.5
Infrared Spectrum Authentic
Specific Rotation +293° to +303° (20°C, 589nm) (c=2, H2O)
Quantity 1 g
Solubility Information Solubility in water: soluble.
Formula Weight 356.37
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Penicillin G sodium salt

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465902 Certificate TypeCertificate of Analysis Date08/11/2024 Catalog Number455690010,455690250,455690050
Lot NumberA0462175 Certificate TypeCertificate of Analysis Date02/07/2024 Catalog Number455690010,455690250,455690050
Lot NumberA0445571 Certificate TypeCertificate of Analysis Date29/04/2024 Catalog Number455690010,455690250,455690050
Lot NumberA0456926 Certificate TypeCertificate of Analysis Date21/11/2023 Catalog Number455690010,455690250,455690050
Lot NumberA0400702 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number455690010,455690250,455690050
compliance-icons
Product Identifier
  • Penicillin G sodium salt
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Penicillin G sodium salt, 96+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.