missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Pectinase from Aspergillus niger
POA
Specifications
Concentration | >1000 U/g |
---|---|
Color | Beige |
For Use With (Application) | Used in plant protoplast preparation to digest cell wall prior to organelle isolation |
Content And Storage | 4°C |
Source | Aspergillus niger |
Description
Pectinase is a macerating enzyme comprised of pectintranseliminase, polygalacturonase, pectinesterase, xylanase, hemicellulases and cellulases.
- Lyophilized
- Activity: >1000 u/gm
Specifications
>1000 U/g | |
Used in plant protoplast preparation to digest cell wall prior to organelle isolation | |
Aspergillus niger | |
Lyophilized Powder |
Beige | |
4°C | |
Pectinase |