Learn More
PD-1/CD279 Mouse anti-Human, CoraLite 647, Clone: 4H4D1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL647-66220-100UL
This item is not returnable.
View return policy
Description
The PD-1/CD279 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PD-1/CD279. This antibody recognizes human, rat, mouse antigen. The PD-1/CD279 antibody has been validated for the following applications: FC, IF analysis. This antibody is coralite™647.
Specifications
PD-1/CD279 | |
Monoclonal | |
2000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Immunofluorescence, Flow Cytometry | |
4H4D1 | |
CoraLite 647 | |
100 μL | |
5133 | |
IgG2b |