missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Path-ID™ qPCR Master Mix
Ambion™ Path-ID™ qPCR Master Mix is a convenient enzyme, buffer, and dNTP mix for TaqMan™ probe-based, real-time PCR detection of DNA targets.
Brand: Applied Biosystems™ 4388643
83.05 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Ambion™ Path-ID™ qPCR Master Mix is optimized for targets of importance in animal health and to function in the presence of PCR inhibitors. The mix is supplied in one tube suitable for 100 reactions. With the exception of PCR primers and probes for your target, the master mix contains everything you need for real-time PCR, including:
- AmpliTaq Gold™ ultrapure thermostable DNA polymerase designed for automatic hot start, to minimize nonspecific PCR products
- Optimized buffer and dNTP concentration, for reliable PCR
- Passive internal reference, based on proprietary ROX™ dye, for precise data analysis.
Specifications
2X qPCR Master Mix is stored at –20°C. Nuclease-free water may be stored at any temperature. | |
Liquid | |
qPCR | |
Fast or Standard | |
ROX (Pre-mixed) | |
qPCR Master Mix | |
Dry Ice | |
Animal Health, Pathogen Detection | |
Built-In Hot Start | |
DNA (Genomic), cDNA |
Primer-probe | |
Tube | |
AmpliTaq Gold DNA Polymerase | |
7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode | |
Ambion™, Path-ID™ | |
UP (Ultra Pure) | |
100 Reactions | |
2X | |
100 Reactions | |
1.375 mL |
For Veterinary Use Only.