missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Parvalbumin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech
This item is not returnable.
View return policy
Description
Parvalbumin Polyclonal antibody apecifically detetects Parvalbumin antigen in Human, Mouse Samples. It is validated for Immunohistochemistry, ELISA.Specifications
Parvalbumin | |
Polyclonal | |
Unconjugated | |
NM_002854 | |
Rabbit | |
Antigen affinity purification | |
Primary | |
Human, Mouse | |
Liquid |
Immunohistochemistry, ELISA | |
1000 μg/mL | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
PVALB | |
Parvalbumin fusion protein Ag30189 | |
150 μL | |
5816 | |
Store at -20°C. Stable for one year after shipment. | |
IgG |