Learn More
Thermo Scientific Chemicals Pararosaniline chloride, pure, high purity biological stain
£21.81 - £398.00
Chemical Identifiers
CAS | 569-61-9 |
---|---|
Molecular Formula | C19H18ClN3 |
Molecular Weight (g/mol) | 323.824 |
MDL Number | MFCD00001657 |
InChI Key | JUQPZRLQQYSMEQ-UHFFFAOYSA-N |
Synonym | Basic Red 9, monohydrochloride, Basic Fuchsin, C.I. 42500 |
PubChem CID | 11292 |
ChEBI | CHEBI:87663 |
IUPAC Name | 4-[(4-aminophenyl)-(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride |
SMILES | C1=CC(=N)C=CC1=C(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10390934
|
Thermo Scientific Chemicals
227880050 |
5 g | Glass bottle |
£21.81
5g |
In Stock
Log in to see stock. |
||||
10104620
|
Thermo Scientific Chemicals
227880250 |
25 g | Glass bottle |
£35.38
25g |
In Stock
Log in to see stock. |
||||
10571671
|
Thermo Scientific Chemicals
227881000 |
100 g | Glass bottle |
£104.52
100g |
In Stock
Log in to see stock. |
||||
11926931
|
Thermo Scientific Chemicals
227885000 |
500 g | Glass bottle |
£398.00
500g |
Estimated Shipment: 16-12-2024
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
569-61-9 | |
323.824 | |
JUQPZRLQQYSMEQ-UHFFFAOYSA-N | |
11292 | |
4-[(4-aminophenyl)-(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride |
C19H18ClN3 | |
MFCD00001657 | |
Basic Red 9, monohydrochloride, Basic Fuchsin, C.I. 42500 | |
CHEBI:87663 | |
C1=CC(=N)C=CC1=C(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N.Cl |
Specifications
268.0°C to 270.0°C | |
C19H18ClN3 | |
Basic Red 9, monohydrochloride, Basic Fuchsin, C.I. 42500 | |
JUQPZRLQQYSMEQ-UHFFFAOYSA-N | |
4-[(4-aminophenyl)-(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride | |
11292 | |
323.83 | |
Pararosaniline chloride |
569-61-9 | |
MFCD00001657 | |
Solubility in water: 10g/L (25°C). Other solubilities: 65g/L ethanol | |
C1=CC(=N)C=CC1=C(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N.Cl | |
323.824 | |
CHEBI:87663 | |
Pure |
Safety and Handling
GHS H Statement:
May cause cancer.
GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 209-321-2
missing translation for 'rtecsNumber' : CX9850100
missing translation for 'tsca' : TSCA
- Pararosaniline chloride
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1B
Hazard Statement
- H350-May cause cancer.
Precautionary Statement
- P201-Obtain special instructions before use.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only