Learn More
Epredia™ Papanicolaou OG-6 and Modified OG Stains
Stain gynecological specimens with Epredia™ Papanicolaou Stain OGs.
£121.00
Products 1
Product Code Brand Type Quantity Price Quantity & Availability  
Product Code Brand Type Quantity Price Quantity & Availability  
10444340
View Documents Promotion Details
Epredia™
75211
OG-6 Papanicolaou Stain 1 gal.
£121.00
3.79 L
Description
Description
  • OG-6 stains the keratinized areas of the cell
  • Modified OG is a Gill modification that minimizes precipitation and reduces staining time
  • Use OG-6 with EA-36 or EA-50 when staining gynecological specimens
  • Use modified OG with EA-65 for staining non-gynecological specimens