missing translation for 'onlineSavingsMsg'
Learn More

Motic™ Panthera TEC POL Digital Microscope

Panthera TEC POL state-of-the-art microscopes are designed for the inspection of all kinds of polarizing materials with birefringence and bireflection.

Brand:  Motic™ 1100403300011

Product Code. 17169368

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Motic™ Panthera TEC POL models present high flexibility for the inspection of all kinds of polarizing materials, transparent samples and opaque specimens, from educational environments to industrial quality control. Even transparent birefringent structures from biology and medicine (heart muscle cells, secondary cell walls, etc.) can be detected by the Full Koehler illumination with exchangeable LED/halogen light source.

  • UC Plan Achromat objectives with focus on strain-free mounting for maximum darkness of image background
  • 3W LED incident light source can easily be changed to different color temperatures
  • Incident/transmitted stands additionally carry Full Koehler illumination with LED/halogen interchangeability
  • 360° rotatable stage for easy positioning of sample
  • Intermediate tube with focusable, centerable Bertrand lens is prepared for conoscopic analysis of crystals and mineral
  • 25° inclined Siedentopf type head with integrated camera
  • Widefield high eyepoint eyepieces UC-WF10X/22 mm with diopter adjustment and rubber eyecups
  • One eyepiece with crosshair reticle and positioning pivot
  • Intermediate body with rotatable analyzer 360°, Bertrand lens and slot for compensators
  • CCIS™ Plan achromatic Strain-free objectives UC 4X/0.1, 10X/0.25, 40X/0.65/S, 60X/0.8/S
  • Motic Light Tracer: Light memory, sleep mode, reversed coded quintuple nosepiece LED light intensity and mode indicator
  • HDMI output, USB for external devices, Ethernet and Wi-Fi
  • Live display mode through Wi-Fi 1920 x 1080 (Full HD), Ethernet 1280 x 720, 1920 x 1080 pixels (Full HD), or HDMI 1920 x 1080 (Full HD)
  • 5 GB internal memory
Specifications

Specifications

Achromat Swing-out Abbe Condenser N.A. 0.90/0.13
CMOS Camera 5MP
Widefield UC-WF10X/22 mm
Koehler LED 3 W and Quartz Halogen 6 V/30 W with Intensity Control
CCIS™ UC Plan Achromatic
Polarizing Microscope
Observing Samples with Birefringence and Bireflection
ImagingOnDevice System (Panthera App Embedded)
0.35 to 30.5 mm
110/240 V
Wi-Fi, HDMI, Ethernet
Live Display
Binocular Siedentopf Type
40X, 100X, 400X, 600X
Panthera TEC POL Epi
5 MP
Blue Filter, Power Cord, Dust Cover, Allen Key, Adjustable Nosepiece Key, Spare HAL Bulb, Interference Color Chart, Screws, 4 Objectives
2 Years Electrical, 5 Years Optical and Mechanical Parts
410 x 300 x 395 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers