missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAIP1 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10675-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene interacts with poly(A)-binding protein and with the cap-binding complex eIF4A. It is involved in translational initiation and protein biosynthesis. Overexpression of this gene in COS7 cells stimulates translation. Alternative splicing occurs at this locus and three transcript variants encoding three distinct isoforms have been identified.Specifications
PAIP1 | |
Polyclonal | |
Unconjugated | |
PAIP1 | |
PABP interacting protein 1, PAIP 1, PAIP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10605, 218693, 365684 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8VE62, Q9H074 | |
Paip1 | |
PAIP1 Fusion Protein Ag1070 | |
150 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |