Learn More
Thermo Scientific™ Oxoid™ PALCAM Selective Supplement
Oxoid PALCAM Selective Supplement is used for the detection of Listeria monocytogenes.
Brand: Thermo Scientific™ SR0150E
Code : 65
Additional Details : Weight : 0.15604kg
Description
Oxoid PALCAM Selective Supplement is used for the detection of Listeria monocytogenes.
Add to PALCAM Agar Base, Part No. CM0877BSpecifications
PALCAM Selective Supplement | |
Selective Supplement | |
Each vial for 500mL medium |
Freeze-dried | |
10/Pk. |