Learn More
Oxalic acid dihydrate, 99+%, extra pure, Thermo Scientific Chemicals
£14.90 - £225.00
Chemical Identifiers
CAS | 6153-56-6 |
---|---|
Molecular Formula | C2H6O6 |
Molecular Weight (g/mol) | 126.064 |
MDL Number | MFCD00149102 |
InChI Key | GEVPUGOOGXGPIO-UHFFFAOYSA-N |
Synonym | oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm |
PubChem CID | 61373 |
IUPAC Name | oxalic acid;dihydrate |
SMILES | C(=O)(C(=O)O)O.O.O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10549243
|
Thermo Scientific Acros
129601000 |
100 g | Plastic Bottle |
£14.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10103280
|
Thermo Scientific Acros
129600010 |
1 kg | Plastic Bottle |
£62.80
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10523314
|
Thermo Scientific Acros
129600050 |
5 kg | Plastic drum |
£225.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6153-56-6 | |
126.064 | |
GEVPUGOOGXGPIO-UHFFFAOYSA-N | |
61373 | |
C(=O)(C(=O)O)O.O.O |
C2H6O6 | |
MFCD00149102 | |
oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm | |
oxalic acid;dihydrate |
Specifications
6153-56-6 | |
99+% | |
HO2CCO2H·2H2O | |
02, 502 | |
15, 7010 | |
Solubility in water: 138g/L (20°C). Other solubilities: 1200g/L in water (90°C), 400g/L in ethanol, 555g/L in boiling alcohol, 181g/L in glycerol, 10g/L in ether | |
C(=O)(C(=O)O)O.O.O | |
126.064 | |
126.04 | |
Extra Pure | |
Low Melting Crystalline Mass, Powder, Crystals and/or Chunks |
98.0°C to 102.0°C | |
C2H6O6 | |
MFCD00149102 | |
01,764; 02,301; 05,481 | |
oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm | |
GEVPUGOOGXGPIO-UHFFFAOYSA-N | |
oxalic acid;dihydrate | |
61373 | |
≥99% | |
Authentic | |
Oxalic acid dihydrate |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Oxalic acid dihydrate
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Oxalic acid dihydrate, Oxalic acid
RUO – Research Use Only