missing translation for 'onlineSavingsMsg'
Learn More

GLASS PLATE FROSTED BLANK 160MM(w) x 140mm (l), 1/8 inch thick

Brand:  Thermo Scientific Owl P1-14FG

GLASS PLATE FROSTED BLANK 160MM(w) x 140mm (l), 1/8 inch thick

Code : 04

Additional Details : Weight : 2.21000kg

Product Code. 12474667

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.