Learn More
Thermo Scientific™ Orion™ Star A Series Portable Meter Conductivity and DO Probe Holder
Use the Thermo Scientific™ Orion™ Star A Series Portable Meter Conductivity and DO Probes Holder with the Star A series portable meter armor to protect your sensors in the field
Brand: Thermo Scientific™ STARA-ESCD
Code : 59
Additional Details : Weight : 1.36080kg
Description
Use the Thermo Scientific™ Orion™ Star A Series Portable Meter Conductivity and DO Probe Holder with the Star A series portable meter armor (Cat. No. STARA-AR) to protect, transport and store your sensors in the field. This conductivity and DO probe holder can be attached to either side of the portable meter armor and holds 12mm diameter or 15mm diameter conductivity probes and a dissolved oxygen probe when placed in its calibration sleeve.
Specifications
Orion Star A series conductivity and dissolved oxygen sensor holder for portable meter protective armor | |
Meter Accessories |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.