Learn More
Thermo Scientific™ Orion™ BNC Meter Shorting Strap
Use the Thermo Scientific™ Orion™ BNC Meter Shorting Strap on pH meters with BNC input to perform BNC input verification testing and meter self test
Brand: Thermo Scientific™ 090045
Code : 59
Additional Details : Weight : 1.00000kg
Description
Use the Thermo Scientific™ Orion™ BNC Meter Shorting Strap on pH meters with BNC input to perform BNC input verification testing and meter self test.
Specifications
Orion BNC shorting cap for pH, ORP and ISE meters | |
Meter Accessories | |
One BNC shorting cap |