missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ AquaMate 7000 Vis Spectrophotometer

Choose the accuracy, dependability and ease of use of the Thermo Scientific™ Orion™ AquaMate 7000 Vis Spectrophotometer to optimize your water and wastewater testing

Brand:  Thermo Scientific™ AQ7000

Product Code. 14865061

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Choose the accuracy, dependability and ease of use of the Thermo Scientific™ Orion™ AquaMate 7000 Vis Spectrophotometer to optimize your water and wastewater testing. This dependable lab spectrophotometer includes 260 preprogrammed methods for ion concentration from absorbance or create custom methods using the standard curve function so virtually any brand of colorimetric reagent can be used. Utilize 325-1100 nm selectable wavelengths, 5 nm spectral bandwidth and vial size flexibility with the included 3-position carousel to hold 13-24 mm round, 10 mm square and 20-50 mm rectangular vials.

Pre-programmed Methods:

  • More than 260 preprogrammed methods offers easy and convenient measurements using Orion AQUAfast™, Merck™ or CHEMetrics™ chemistries
  • Preprogrammed methods can also be adapted for new chemistries or operators can create their own custom methods and calibration curves
  • Instruments allow a one-point adjustment on any preprogrammed method to correct for variations in batch reagent chemistries

Versatility:

  • Wide variety of vials sizes can be used, including 10mm square vials, 20 and 50mm rectangular vials and an adjustable round vial holder that accommodates 13, 16, and 24mm round vials
  • Three USB connections provide the options to store methods and data to a USB memory stick, transfer data to a computer and print hard copy data to an external printer
  • System is compatible with ink jet and laser printers with HP PCL 5 or greater

Verification:

  • Performance verification tests ensure wavelength accuracy and instrument functionality
  • Each verification report gives the time, date and instrument serial number in accordance with GLP and GMP
  • Built-in wavelength accuracy test is compatible with either the internal lamp or external calibrated standards
  • Built-in test methods for stray light, noise and resolution are available to validate instrument performance further

Includes:

  • Methods on USB memory stick
  • Three-position cell holder
  • User documentation
  • Dust cover
  • USB cable
  • 110V, 220V and 240V power cords
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Compatible with ink jet and laser printers with HP PCL 5 or greater
AquaMate 7000 Vis Spectrophotometer
Graphical with LCD backlight, 9.7 x 7.1 cm (3.8 x 2.8 in.)
5 nm
325 to 1100 nm
100/240 V, 50/60 Hz
25 cm
Methods on USB memory stick; 3-position cell older; User documentation; Dust cover; USB cable 110V and 220V power cords
1000 hrs.
Single Beam
-0.1 to 3.0 A ; -0.3 to 125 %T ; ±9999 C
10 to 4200 nm/min.
±1 nm
±0.5nm
19 lb.
11.8 in.
USB type A port for USB stick (front panel)
USB type B port for computer (rear panel)
USB type A port for printer (rear panel)
Silicon Photodiode
Tungsten Halogen
100/240 V
0.002 A/hr. after warmup
9.8 in.
50/60 Hz
Sealed membrane with tactile response keys
<0.001 A at 0.0 A; <0.002 A at 2.0 A Peak-to peak at 340nm
Up to 3.0A at 340nm
External Printer: Ink Jet, Laser Printer with HP PCL 5 or Greater
<0.08 %T at 220 nm and 340 nm; <0.1 %T at 340 nm and 400 nm; <1.0 %T at 198 nm
1.0; 2.0; 3.0; 5.0 nm
11,000 nm/min.
8.6 kg
30 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers