Learn More
Invitrogen™ OPG (TNFRSF11B) Mouse ELISA Kit
Sandwich ELISA Kit
£504.00 - £2320.00
Specifications
Accession Number | O08712 |
---|---|
Assay Sensitivity | 1 pg/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15405343
|
Invitrogen™
EMTNFRSF11B |
96 Tests |
£504.00
1 set |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
17115789
|
Invitrogen™
EMTNFRSF11BX5 |
5 x 96 Tests |
£2320.00
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
Description
The Mouse Osteoprotegerin (OPG/TNFRSF11B) ELISA quantitates Ms OPG in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms OPG. Principle of the method The Mouse OPG solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Acts as decoy receptor for TNFSF11/RANKL and thereby neutralizes its function in osteoclastogenesis. Inhibits the activation of osteoclasts and promotes osteoclast apoptosis in vitro. Bone homeostasis seems to depend on the local ratio between TNFSF11 and TNFRSF11B. May also play a role in preventing arterial calcification. May act as decoy receptor for TNFSF10/TRAIL and protect against apoptosis. TNFSF10/TRAIL binding blocks the inhibition of osteoclastogenesis.Specifications
O08712 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin , Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN- gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-3 Rb, IL-4, IL-5, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP- 5, M-CSF, MIG, MIP-1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, PSelectin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF-alpha, TNF RI, TNF RII, TPO, VCAM-1, VEGF. | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
18383 | |
<12% | |
HRP | |
Apoptosis | |
OCIF, Opg, TR1 | |
1 hr. 20 min. |
1 pg/mL | |
Biotin | |
Other Proteins | |
Mouse | |
Colorimetric Microplate Reader | |
OCIF,Opg,TR1 | |
<10% | |
RUO | |
2°C to 8°C | |
Mouse | |
4 hr. 45 min. |